missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Solvent FI, Honeywell Fluka™

Medium for volumetric, two-component Karl Fischer titrations

Brand:  Honeywell Fluka 34432-6X1L


 View more versions of this product

Product Code. 16812370

  • £394.33 / 6 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Methanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - NEXTGEN Solvent FI
11°C
147-85-3
≥0.10%
Glass Bottle
UN1230
0.9 g/cm3 (at 20°C)
6 x 1 L
5 to 6
≤5%
Liquid
Completely Miscible in Water
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    7

    Hours

    5

    Minutes

    10

    Seconds

    30
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - NEXTGEN Solvent FI, Honeywell Fluka™ > 6 x 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.