Learn More
Hydrazine dihydrochloride, 98+%, Thermo Scientific Chemicals
Description
Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 200°C (decomposition) |
Melting Point | 198°C |
Color | White |
Physical Form | Lump |
Quantity | 500 g |
Assay Percent Range | >98% |
Solubility Information | Soluble in water. Slightly soluble in alcohol |
Formula Weight | 104.97 |
Odor | Odorless |
Appearance | White crystalline or lump |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X16K041 | Certificate of Analysis | 30/10/2024 | 032725.36,032725.22 |
U01K044 | Certificate of Analysis | 16/08/2024 | 032725.36,032725.22 |
U09K062 | Certificate of Analysis | 14/08/2024 | 032725.36,032725.22 |
R06K005 | Certificate of Analysis | 15/05/2024 | 032725.36,032725.22 |
M16K013 | Certificate of Analysis | 23/01/2024 | 032725.36,032725.22 |
Safety and Handling



- Hydrazine dihydrochloride
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin sensitiser Category 1
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.