Learn More
Hydrazine dihydrochloride, 98+%, Thermo Scientific Chemicals
For Use As Chlorine Scavenger in Hcl Gas Streams
£48.94 - £176.88
Chemical Identifiers
CAS | 5341-61-7 |
---|---|
Molecular Formula | ClH5N2 |
Molecular Weight (g/mol) | 68.50 |
MDL Number | MFCD00064543 |
InChI Key | BIVUUOPIAYRCAP-UHFFFAOYSA-N |
Synonym | hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl |
PubChem CID | 17548 |
IUPAC Name | hydrazine;dihydrochloride |
SMILES | [H+].[Cl-].NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11332988
|
Thermo Scientific Alfa Aesar
032725.22 |
100 g |
£48.94
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342988
|
Thermo Scientific Alfa Aesar
032725.36 |
500 g |
£176.88
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5341-61-7 | |
68.50 | |
BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
17548 | |
[H+].[Cl-].NN |
ClH5N2 | |
MFCD00064543 | |
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl | |
hydrazine;dihydrochloride |
Specifications
200°C (decomposition) | |
5341-61-7 | |
Lump | |
ClH5N2 | |
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl | |
BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
hydrazine;dihydrochloride | |
17548 | |
Odorless | |
1.42g/cm3 |
198°C | |
White | |
>98% | |
MFCD00064543 | |
Soluble in water. Slightly soluble in alcohol | |
[H+].[Cl-].NN | |
68.50 | |
104.97 | |
White crystalline or lump | |
Hydrazine dihydrochloride, Special handling precautions required |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H350
missing translation for 'dotInformation' : DOT Class: 6.1, Packing Group: III
missing translation for 'einecsNumber' : 226-283-2
missing translation for 'rtecsNumber' : MV2298000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Hydrazine dihydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin sensitiser Category 1
Hazard Statement
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only