missing translation for 'onlineSavingsMsg'
Learn More
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals Product Code.: 10646594

Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals

Product Code. 10646594
1 kg, Glass bottle
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10646594

Brand: Thermo Scientific 325940010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2950-43-8
Molecular Formula H3NO4S
Molecular Weight (g/mol) 113.09
MDL Number MFCD00011604
InChI Key DQPBABKTKYNPMH-UHFFFAOYSA-N
Synonym hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxideShow MoreShow Less
PubChem CID 76284
IUPAC Name amino hydrogen sulfate
SMILES NOS(O)(=O)=O

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula H2NOSO3H
Quantity 1 kg
Fieser 01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17
Solubility Information Solubility in water: soluble
Formula Weight 113.09
Percent Purity 0.97
Physical Form Crystalline Powder
Chemical Name or Material Hydroxylamine-O-sulfonic acid, 0.97
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0413764 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number325940250,325940010,325941000
Lot NumberA0406589 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number325940250,325940010,325941000
Lot NumberA0447627 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number325940250,325940010,325941000
Lot NumberA0442848 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog Number325940250,325940010,325941000
Lot NumberA0436779 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number325940250,325940010,325941000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine-O-sulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.