Learn More
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
£46.33 - £857.27
Chemical Identifiers
CAS | 2950-43-8 |
---|---|
Molecular Formula | H3NO4S |
Molecular Weight (g/mol) | 113.09 |
MDL Number | MFCD00011604 |
InChI Key | DQPBABKTKYNPMH-UHFFFAOYSA-N |
Synonym | hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxideShow More |
PubChem CID | 76284 |
IUPAC Name | amino hydrogen sulfate |
SMILES | NOS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10428662
|
Thermo Scientific Acros
325940250 |
25 g | Glass bottle |
£46.33
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10692322
|
Thermo Scientific Acros
325941000 |
100 g | Glass bottle |
£125.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646594
|
Thermo Scientific
325940010 |
1 kg | Glass bottle |
£857.27
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2950-43-8 | |
113.09 | |
DQPBABKTKYNPMH-UHFFFAOYSA-N | |
76284 | |
NOS(O)(=O)=O |
H3NO4S | |
MFCD00011604 | |
hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxideShow More | |
amino hydrogen sulfate |
Specifications
2950-43-8 | |
White | |
0.97 | |
H2NOSO3H | |
01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17 | |
Solubility in water: soluble | |
NOS(O)(=O)=O | |
113.09 | |
113.09 | |
Crystalline Powder |
210.0°C | |
Authentic | |
H3NO4S | |
MFCD00011604 | |
hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxide | |
DQPBABKTKYNPMH-UHFFFAOYSA-N | |
amino hydrogen sulfate | |
76284 | |
0.97 | |
Hydroxylamine-O-sulfonic acid, 0.97 |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-971-6
missing translation for 'rtecsNumber' : NC5427000
missing translation for 'tsca' : TSCA
- Hydroxylamine-O-sulfonic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.