Learn More
Invitrogen™ Silencer™ Select Negative Control No. 1 siRNA
A non-targeting negative control siRNA with the same chemical modifications for enhanced efficacy as in other Silencer™ Select siRNAs.
Brand:  Invitrogen™ 4390843
 View more versions of this product
Product Code. 10025994
  • £244.00 / Each
Edge
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • A non-targeting negative control siRNA with the same chemical modifications for enhanced efficacy as in other Silencer™ Select siRNAs
  • Validated siRNA controls for optimizing siRNA experiments
  • GAPDH Positive Control functionally tested in several common cell lines
  • Negative Controls functionally proven to have minimal effects on cell proliferation and viability
  • Include Silencer™ Select modifications for enhanced specificity
  • For use in human, mouse, and rat cells
  • Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C
What are Silencer™ Select siRNAs?
  • Silencer™Select siRNAs are our next-generation siRNAs
  • These siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications to yield siRNAs with unrivalled efficacy, potency, and specificity
  • The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data
Silencer™ Select Negative Control siRNAs
  • Negative control siRNAs are siRNAs with sequences that do not target any gene product
  • They are essential for determining the effects of siRNA delivery and for providing a baseline to compare siRNA-treated samples
  • These negative control siRNAs have been tested by microarray analysis and shown to have minimal effects on gene expression
  • In addition, these two controls have been tested in multi-parametric, cell-based assays and are proven to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains dried siRNA at (5 nmol) and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °.
siRNA
Unconjugated
siRNA
5 nmol
Negative Control
RNAi, Transfection
Silencer™, Silencer™ Select, Ambion™
HPLC
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Silencer™ Select Negative Control No. 1 siRNA > 5 nmol