Learn More
Ibuprofen, 99%, Thermo Scientific Chemicals
Ibuprofen, 2-4-isobutylphenyl propanoic acid, CAS # 15687-27-1, is a laboratory form of the over-the-counter nonsteroidal anti-inflammatory (NSAID) drug.
£44.50 - £887.00
Chemical Identifiers
CAS | 15687-27-1 |
---|---|
Molecular Formula | C13H18O2 |
Molecular Weight (g/mol) | 206.29 |
MDL Number | MFCD00010393 |
InChI Key | HEFNNWSXXWATRW-UHFFFAOYNA-N |
IUPAC Name | 2-[4-(2-methylpropyl)phenyl]propanoic acid |
SMILES | CC(C)CC1=CC=C(C=C1)C(C)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15853855
|
Thermo Scientific Acros
258610010 |
1 g | Glass bottle |
£44.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10768501
|
Thermo Scientific Acros
258610100 |
10 g | Glass bottle |
£232.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964991
|
Thermo Scientific Acros
258610500 |
50 g | Glass bottle |
£887.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Ibuprofen (2-4-isobutylphenyl propanoic acid) is a common NSAID that reduces pyrogen release and inflammation
- Ibuprofen is a racemic mixture that consists of an R-enantiomer and S-enantiomer; the S-enantiomer is proposed to be the stronger acting agent
- It is a propionic acid derivative, a short-chain saturated fatty acid bound to ethane
- Ibuprofen inhibits cyclooxygenases and reduces prostaglandin formation, inhibiting fever and pain
Applications
- Ibuprofen reduces both local and systemic inflammation; suitable for use in vitro or in vivo to study specific or broad mechanisms
- Intended for sensitive biochemical applications such as HPLC
Chemical Identifiers
15687-27-1 | |
206.29 | |
HEFNNWSXXWATRW-UHFFFAOYNA-N | |
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O |
C13H18O2 | |
MFCD00010393 | |
2-[4-(2-methylpropyl)phenyl]propanoic acid |
Specifications
15687-27-1 | |
Authentic | |
C13H18O2 | |
MFCD00010393 | |
HEFNNWSXXWATRW-UHFFFAOYNA-N | |
2-[4-(2-methylpropyl)phenyl]propanoic acid | |
206.27 | |
Crystalline Powder |
75°C to 78°C | |
99% | |
(CH3)2CHCH2C6H4CH(CH3)COOH | |
α-Methyl-4-(2-methylpropyl)benzeneacetic acid, p-Isobutylhydratropic acid, 2-(4-Isobutylphenyl)-propionic acid | |
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O | |
206.29 | |
98.5% min. | |
Ibuprofen |
Safety and Handling
GHS P Statement Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only