missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals IKK-2 Inhibitor V Product Code.: 15436749

Thermo Scientific Chemicals IKK-2 Inhibitor V

Product Code. 15436749
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 15436749

Brand: Thermo Scientific Chemicals J65132.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

A selective IKK-2 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A selective IKK-2 inhibitor
CAS 978-62-1
Molecular Formula C15H8ClF6NO2, C15H8ClF6NO2
Formula Weight 383.67g/mol
Synonym N-(3,5-Bis-trifluoromethylphenyl)-5-chloro-2-hydroxybenzamide; IMD-0354
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Powder
InChI Key CHILCFMQWMQVAL-UHFFFAOYSA-N
SMILES OC1=C(C=C(Cl)C=C1)C(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
Molecular Weight (g/mol) 383.67
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25I001 Certificate TypeCertificate of Analysis Date01/11/2022 Catalog NumberJ65132.MA
Lot NumberM12C008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ65132.MA
Lot NumberB26Z009 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ65132.MA
Lot NumberS15D035 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ65132.MA
Lot NumberT06B014 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ65132.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals IKK-2 Inhibitor V >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.