missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications. Cytiva Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.
£142.50 - £645.00
Specifications
Content And Storage | Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP |
---|---|
Quantity | 1 Kit |
For Use With (Application) | Standard PCR |
Product Code | Brand | No. of Reactions | Volume | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Volume | Price | Quantity & Availability | ||||
10525775
|
Cytiva
27955702 |
480 | 200μL |
£645.00
480 reactions |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10380395
|
Cytiva
27-9557-01 |
96 | 200μL |
£142.50
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10678095
|
Cytiva
27-9559-01 |
96 | 200μL |
£144.81
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10289034
|
Cytiva
27-9558-01 |
100 | 500μL |
£156.00
100 reactions |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
Specifications
Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP | |
1 Kit | |
Standard PCR |