missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ BSA (in MES Buffer)

Easily prepare immunogens from carboxyl-containing peptide antigens with bovine serum albumin (BSA) and different conjugation chemistries.

Brand:  Thermo Scientific™ 77171

43.83 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.



Product Code. 11894071

  • £65.75 / 2mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

4°C
Imject Bovine Serum Albumin (in MES buffer)
2 mg
Imject™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK409994 Certificate TypeCertificate of Analysis Date18/12/2024 Catalog Number77171
Lot NumberZK409482 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number77171
Lot NumberYL379577 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77171
Lot NumberYF372936 Certificate TypeCertificate of Analysis Date06/07/2023 Catalog Number77171
Lot NumberYC365272 Certificate TypeCertificate of Analysis Date16/03/2023 Catalog Number77171
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ BSA (in MES Buffer) > BSA in MES buffer; 2mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.