Learn More
Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells, with a controlled, contamination-free atmosphere in each culture tray.
Brand: Thermo Scientific™ 173238
Description
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells using Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing. Flow system helps ensure a controlled, contamination-free atmosphere in each culture tray by equally distributing the user-specified gas mixture. Available in 4-, 10- and 40-tray models.
- Achieve greater yields with controlled cell culture environments equally distributed within each layer
- Optimize production capacity without changing cell culture platforms
- Develop industrial scale solutions with the utilization of an active gassing Cell Factory™ Incubator
Specifications
Cell Factory with Active Gassing | |
6 | |
Sterile | |
10 |
6 | |
System with Active Gassing | |
6320 cm² | |
Pack/Case: 1/6 |
Product Suggestions
Customers who viewed this item also viewed.