missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CELLSTACK™ Culture Chambers
GREENER_CHOICEBIOPHARMA

Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.

Brand:  Corning™ 3269

355.91 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10377862

  • £396.41 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
  • Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
  • Sterilized by gamma irradiation
  • Leak tested

  • Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
  • Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation

Order Info

Individually packaged

Certifications

Certified nonpyrogenic.

Compliance

Polystyrene material meets USP Class VI requirements; provides superior optical clarity and structural integrity.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8.45 to 12.84 oz.
Orange
250 to 380 mL
Culture Chamber
1272 cm²
Two chambers
USP Class VI
33mm
Polystyrene
Yes
Tissue Culture-treated
Gamma Irradiated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number29524004 Certificate TypeCertificate of Quality Date11/04/2025 Catalog Number3269
Lot Number27324014 Certificate TypeCertificate of Quality Date25/03/2025 Catalog Number3269
Lot Number25224004 Certificate TypeCertificate of Quality Date12/02/2025 Catalog Number3269
Lot Number23724005 Certificate TypeCertificate of Quality Date21/12/2024 Catalog Number3269
Lot Number22824028 Certificate TypeCertificate of Quality Date18/12/2024 Catalog Number3269
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    18

    Hours

    5

    Minutes

    9

    Seconds

    25
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CELLSTACK™ Culture Chambers > Tissue Culture Surface; 2 Chambers; Total Culture Area: 1,272 cm2; 5/Cs.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.