missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Freund's Complete Adjuvant (FCA)

Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.

Brand:  Thermo Scientific™ 77140

129.20 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Product Code. 11824071

  • £139.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Imject Freund's Adjuvants are 10mL ampules of complete (FCA) and incomplete (FIA) formulations of a popular nonspecific stimulator of the immune response for use in antibody production immunization protocols.

The water-and-oil emulsions (with and without inactivated Mycobacterium) greatly enhance immune responses to immunogens when mixed and injected with the prepared antigen. Typically, FCA is used for initial injections and FIA is used for booster injections. The ampules comprise conveniently sized aliquots that provide long shelf life.

Highlights:

  • Water-in-oil emulsion with or without killed Mycobacterium
  • Enhance immune response and antibody production for injected immunogens
  • Useful for immunization of mice, rabbits and other mammalian hosts
  • Requires emulsification with prepared immunogens

Freund's Complete Adjuvant (FCA), Part No. 77140:

  • Water-in-oil emulsion containing killed Mycobacterium
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Disadvantages: difficult to mix with immunogen and can cause tissue necrosis at injection site

Freund's Incomplete Adjuvant (FIA), Part No. 77145:

  • Water-in-oil emulsion without Mycobacterium
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Disadvantage: difficult to mix with immunogen
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Upon receipt store at 4°C
Imject Freund's Complete Adjuvant (FCA)
5 x 10 mL
Imject™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZD386629 Certificate TypeCertificate of Analysis Date20/04/2024 Catalog Number77140
Lot NumberZD387194 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number77140
Lot NumberZC385234 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog Number77140
Lot NumberZA384661 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number77140
Lot NumberYJ378674 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number77140
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ Freund's Complete Adjuvant (FCA) > Complete Adjuvant (FCA); 5 x 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.