missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Maleimide-Activated BSA Spin Kit

Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.

Brand:  Thermo Scientific™ 77667

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11874035

  • £610.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient For: 5 conjugation reactions and complete immunogen preparations, each with up to 2 mg sulfhydryl-containing peptide
• Imject Maleimide-Activated BSA, 5 x 2 mg
• Imject Maleimide Conjugation Buffer, 30 mL
• Imject Purification Buffer Salts, 5 x 0.84 g
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns

Store the Maleimide-Activated blue Carrier Protein at -20°C. Store remaining components at 4°C.
Kit
Imject™
Imject Maleimide-Activated BSA Spin Kit
5 x 2 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ394409 Certificate TypeCertificate of Analysis Date09/11/2024 Catalog Number77667
Lot NumberZA386248 Certificate TypeCertificate of Analysis Date03/02/2024 Catalog Number77667
Lot NumberYI380530 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog Number77667
Lot NumberYF363459 Certificate TypeCertificate of Analysis Date22/06/2023 Catalog Number77667
Lot NumberSPECS Certificate TypeCertificate of Analysis Date28/10/2022 Catalog Number77667
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ Maleimide-Activated BSA Spin Kit > Maleimide Conjugation Kit; 5 x 2mg kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.