missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Brand:  Thermo Scientific™ 343036

 View more versions of this product

Product Code. 10172831

  • POA / Pack of 2400

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Polyethylene
15 mm
2400
15 mm Nunc-Immuno Tubes
Stopper
300
Not Applicable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184539 Certificate TypeCertificate of Compliance Date04/02/2025 Catalog Number343036
Lot Number170599 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number343036
Lot Number179044 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number343036
Lot Number182889 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number343036
Lot Number177163 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number343036
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Tubes and Stoppers > Nunc-Immuno, 15

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.