Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand:  Fisherbrand™ FB67811
 View more versions of this product
Product Code. 15505099
  • £43.06 / 1 unit
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class E2 calibration weight
  • In compliance with OIML R111
Specifications
Specifications
E2
Stainless Steel
500 mg
500 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2 > 500mg