missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 51.0°C to 53.0°C |
Color | Pink to White |
Density | 1.2200g/mL |
Boiling Point | 253.0°C to 254.0°C |
Flash Point | 136°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Molecular Formula | C8H7N |
Quantity | 25 g |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466521 | Certificate of Analysis | 08/10/2024 | 122150025,122150250,122150100,122155000,122151000 |
A0465795 | Certificate of Analysis | 04/09/2024 | 122150025,122150250,122150100,122155000,122151000 |
A0458653 | Certificate of Analysis | 13/12/2023 | 122150025,122150250,122150100,122155000,122151000 |
A0429310 | Certificate of Analysis | 20/10/2023 | 122150025,122150250,122150100,122155000,122151000 |
A0453025 | Certificate of Analysis | 25/08/2023 | 122150025,122150250,122150100,122155000,122151000 |
Safety and Handling


Product Identifier
- Indole
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Indole
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Indole, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction