Learn More
3M™ Industrial Headgear Combination
3M™ Industrial Headgear Combination provides a versatile and comfortable solution for wearers who require both face and hearing protection.
Brand: 3M™ 7100029146
562.52 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- A practical, convenient solution for face and hearing protection where no head protection is needed
- Front cover provides ventilation for increased comfort and also designed as a cable holder for combination with communication products
- Clear Face Shield: Polycarbonate is approved for High Speed Particles 190 m/s according to EN166:2001
- To increase eye protection, G500 can incorporate the 3M™ V6 integrated safety glasses range, that are available in clear, gray and yellow lenses
- Headgear can be used both with and without earmuffs
- Headband is slotted for the attachment of earmuffs
Specifications
Yellow | |
Headgear Combination |