Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Brand:  Mettler Toledo™ 30281917
100.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15589651
  • £106.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S7-BNC
To connect Mettler Toledo InLab sensors to any meter
5.0 m
Mettler Toledo InLab Sensors
16.4 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection > Connector: BNC; Length: 5m (16.4 ft.)