Learn More
INSR alpha Monoclonal Antibody (83-14), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHR0221
Description
This antibody reacts with an epitope at aa 469-592 (exon 7/8). It primarily reacts with human, but also reacts very weakly with cow, pig and sheep. This antibody can inhibit insulin binding (∽80%). It can also be used in a Tyrosine Kinase assay (Ab-mediated capture on microtitre plates). Without BSA, this antibody can be used as an insulin-like agonist. Without BSA, it can also be used as both a capture and detection antibody in a sandwich ELISA.
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
Specifications
INSR alpha | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P06213 | |
INSR | |
IM-9 lymphocytes followed by purified insulin receptor. | |
500 μL | |
Primary | |
Bovine, Human, Ovine, Pig | |
Antibody | |
IgG2a κ |
ELISA, Functional Assay, Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Western Blot | |
83-14 | |
Unconjugated | |
INSR | |
4932439J01Rik; CD220; CD220 beta; D630014A15Rik; DAF-2; HHF5; INSR; insulin receptor; insulin receptor (INSR); insulin receptor precursor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR alpha; IR beta; ir-A; IR-B | |
Mouse | |
Protein A | |
RUO | |
3643, 396755, 408017, 443431 | |
4° C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.