Learn More
Invitrogen™ AccuPrime™ Pfx DNA Polymerase
AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis.
Brand: Invitrogen™ 12344024
381.35 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: AccuPrime Pfx DNA Polymerase (100μL, at 2.5U/μL) is provided with 10X AccuPrime reaction mix (1mL) that contains dNTPs and a separate vial (1mL) of 50mM MgSO4. Reagents are provided for 200 amplification reactions of 50μL each.
Description
AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading (3'→5' exonuclease) activity. Platinum anti-Pfx DNA polymerase antibodies inhibit polymerase activity, providing hot-start capabilities for improved PCR specificity (see figure). Thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, increasing specificity, yield, and robustness over Platinum Pfx alone.
Benefits of AccuPrime Pfx DNA Polymerase
- Greater than 26 times higher fidelity than Taq DNA polymerase
- Amplification of fragments up to 12 kb
- Minimized PCR optimization
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- RT-PCR
- High-fidelity PCR
- Hot-start PCR
Notes
- AccuPrime Pfx DNA Polymerase is especially effective where very high levels of fidelity are required.
- The 10X AccuPrime Pfx reaction mix contains MgSO4, dNTPs, and thermostable proteins that enhance specific primer-template hybridization during each PCR cycle.
Order Info
Approved for shipment on Wet or Dry Ice
Specifications
• AccuPrime Pfx DNA Polymerase (1 x 100 μL at 2.5 U/μL) • 10X AccuPrime Pfx Reaction Mix (1 x 1 mL) • 50 mM MgSO4 (1 x 1 mL) Store at -20°C. |
|
Low | |
Standard | |
Wet or Dry Ice | |
26X | |
200 Reactions | |
Separate Components | |
100 μL |
Primer-probe | |
AccuPrime Pfx DNA Polymerase | |
200 Reactions | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
Blunt | |
12 kb or less |
For Research Use Only. Not for use in diagnostic procedures.