missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anchored Oligo(dT)20 Primer

Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC).

Brand:  Invitrogen™ 12577011

Product Code. 11662244

  • £170.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC). Due to the variable end-sequence, the primer is anchored to the 5´-end of the poly(A) tail of mRNA and prevents priming within the poly(A) tail.

Anchored Oligo(dT)20 Primer is recommended for use in cDNA labeling protocols and in RT-PCR. It is commonly used for first-strand cDNA systhesis.

Quality control: This product is qualified in a first-strand cDNA synthesis reaction

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Primer
Phosphate
5'd PO4 [(T)20VN]3'
Dry Ice
Anchored Oligo dT Primer
First-strand cDNA Synthesis
• Anchored Oligo(dT)20 Primer in DEPC-treated water (20 μL at 2.5 μg/μL)

Store at –20°C.
Guaranteed stable for 6 months when properly stored.
22-mer
Gel-purified, Desalted
2.5 μg/μL
20 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Anchored Oligo(dT)20 Primer > 50 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.