Learn More
Invitrogen™ Dynabeads™ MyOne™ Carboxylic Acid
Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups.
Brand: Invitrogen™ 65011
120.65 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups. Rapid binding chemistry, activation through carbodimide is required. Amino-modified oligonucleotides, antibodies or other ligands can easily be coupled to the beads. Low non-specific binding of nucleic acids. The 1 μm Dynabeads™ MyOne™ have a large surface area, high capacity, efficient magnetic pull and a slow sedimentation rate during incubation. The MyOne™ products are tailor-made for use in automated protocols where high throughput is crucial.
Benefits and Features:
• Ideal for use as a solid support in in vitro diagnostic assays.
• Capture of specific DNA/RNA sequences, proteins or any other analyte.
• High magnetic mobility and low sedimentation rate
Concentration:
10 mg/mL
Order Info
Shipping Condition: Room Temperature
Specifications
Store in refrigerator (2–8°C). | |
5 μL | |
Room Temperature | |
Carboxylic Acid | |
2 mL | |
Carboxylic Acid Magnetic Bead | |
Research Grade |
1 μm | |
High-throughput Compatible | |
Any Sample Type | |
Dynabeads™, MyOne™ | |
Protein Purification and Isolation | |
Beads in solution |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.