Learn More
Invitrogen™ Dynabeads™ MyOne™ Carboxylic Acid
Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups.
£132.00 - £471.00
Specifications
Content And Storage | Store in refrigerator (2–8°C). |
---|---|
Diameter (Metric) | 1 μm |
Elution Volume Min | 5 μL |
High-throughput Compatibility | High-throughput Compatible |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10563775
|
Invitrogen™
65011 |
2 mL |
£132.00
Each |
In Stock
Log in to see stock. |
|||||
10503375
|
Invitrogen™
65012 |
10 mL |
£471.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups. Rapid binding chemistry, activation through carbodimide is required. Amino-modified oligonucleotides, antibodies or other ligands can easily be coupled to the beads. Low non-specific binding of nucleic acids. The 1 μm Dynabeads™ MyOne™ have a large surface area, high capacity, efficient magnetic pull and a slow sedimentation rate during incubation. The MyOne™ products are tailor-made for use in automated protocols where high throughput is crucial.
Benefits and Features:
• Ideal for use as a solid support in in vitro diagnostic assays.
• Capture of specific DNA/RNA sequences, proteins or any other analyte.
• High magnetic mobility and low sedimentation rate
Concentration:
10 mg/mL
Order Info
Shipping Condition: Room Temperature
Specifications
Store in refrigerator (2–8°C). | |
5 μL | |
Room Temperature | |
Carboxylic Acid | |
Protein Purification and Isolation | |
Beads in solution |
1 μm | |
High-throughput Compatible | |
Any Sample Type | |
Dynabeads™, MyOne™ | |
Carboxylic Acid Magnetic Bead | |
Research Grade |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.