missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flp-In™-3T3 Cell Line

Designed for rapid generation of stable cell lines that express protein of interest from Flp-In expression vector

Brand:  Invitrogen™ R76107

1623.60 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10286832

  • £1804.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 3 x 106 cells are supplied frozen in 1mL 90% complete medium and 10% DMSO.
This item is not returnable. View return policy

Description

Description

  • These cells contain single stably integrated FRT site at transcriptionally active genomic locus
  • Targeted integration of Flp-Inexpression vector ensures high-level expression of gene of interest
  • Created by transfecting parent cell lines with pFRT/lacZeo and selecting for stable Zeocin™-resistant clones
  • Flp-In-3T3 cells tend to down regulate CMV promoter

Constitutive Expression, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 x 106 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.
Mouse
1 mL
Flp-In™-3T3
pSec
Flp-In™
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Flp-In™-3T3 Cell Line > 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.