Learn More
Invitrogen™ Lipofectin™ Transfection Reagent
Ideal reagent for transfection of endothelial cells
£512.00 - £1532.00
Specifications
Cell Type | Established Cell Lines, Primary Cells, Hard-to-Transfect Cells |
---|---|
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Plasmid DNA |
Transfection Technique | Lipid-based Transfection |
For Use With (Application) | Transfection |
Includes: Contains one vial (1mL) of Lipofectin Transfection Reagent.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10104432
|
Invitrogen™
18292037 |
4 mL |
£1532.00
4mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10685773
|
Invitrogen™
18292011 |
1 mL |
£512.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For transfecting DNA, RNA, and oligonucleotides into mammalian cells, and DNA into plant protoplasts
- Lipofectin reagent has also been shown to work well, in combination with PLUS™ Reagent, for transfection of HeLa cells
- Lipofectin Transfection Reagent is a 1:1 (w/w) liposome formulation of cationic lipid N-[1-(2,3-dioleyloxy)propyl]-n,n,n-trimethylammonium chloride (DOTMA) and dioleoyl phophotidylethanolamine (DOPE) in membrane-filtered water
- Animal-free origin
Drug Discovery & Development, Gene Expression Analysis & Genotyping, PCR & Real-Time PCR, PCR-Based Drug Discovery Assays, Real Time PCR (qPCR), Real Time PCR-Based Gene Expression Profiling, Target & Lead Identification & Validation
Order Info
Shipping Condition: Wet Ice
Specifications
Established Cell Lines, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA | |
Transfection | |
Lipofectin™ | |
No |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
Not High-throughput Compatible (Manual) | |
Transfection Reagent | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.