missing translation for 'onlineSavingsMsg'
Learn More

Phasemaker™ Phasemaker™ Tubes

Invitrogen™ Phasemaker™ Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol™ mix from the organic phase underneath, making it very easy to pipette off the upper phase containing the RNA.

Brand:  Phasemaker™ A33248

84.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15645268

  • £93.50 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: One bag of 100 Phasemaker Tubes
This item is not returnable. View return policy

Description

Description

Invitrogen™ Phasemaker™ Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol™ mix from the organic phase underneath, making it very easy to pipette off the upper phase containing the RNA. The polymer works on a density gradient, as it is heavier than the aqueous phase, but lighter than the organic phase; after centrifugation it positions itself between these two layers. It creates a tight seal between the two phases, making it very easy to pipette off the aqueous phase without  the risk of carrying over contaminants from the organic layer. Because this, Invitrogen Phasemaker Tubes allow recovery of the whole aqueous phase, increasing yield up to 30%. It is an inert polymer that does not interfere with downstream experiments such as PCR. It works for RNA extraction using TRIzol™, TRIzol™ LS and TRIzol™ Plus only. It is not recommended to be used with other organic reagents as their density might be different. It works with fresh or frozen samples (tissue or blood). It also  works with tissue samples treated previously with RNAlater. Invitrogen Phasemaker Tubes come already pre-dispensed. The tube size is  2 mL.

  • Pre-dispensed in 2 mL tubes
  • Creates a tight seal between the aqueous and the organic phases
  • Easier to pipette off the whole aqueous phase
  • Increases yield up to 30%
  • RNA integrity is not compromised
  • The seal stays for longer periods of time, allowing you to increase the number of samples per batch
  • Inert, does not interfere with downstream experiments

Services

One year after receiving

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 Phasemaker Tubes; room temperature

Total RNA
100 Tubes
Cells: ≤107
Tissue: ≤100 mg
Cells, Tissue
Not High-throughput Compatible (Manual)
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number563011655 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA33248
Lot Number57206733 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA33248
Lot Number5750210 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA33248
Lot Number57502391 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA33248
Lot Number57502668 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA33248
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phasemaker™ Phasemaker™ Tubes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.