Learn More
Invitrogen™ Qubit™ 4 Starter Packages, with WiFi
Accurately quantify protein, dsDNA, and RNA with the Qubit 4 Protein BR, Quantitation, NGS, and RNA IQ Starter Kits, which feature a fluorescent dye that emits signal only when bound to its target molecule, minimizing the effect of contaminants.
Brand: Invitrogen™ Q33240
2353.05 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
Achieve accurate and reliable results with Qubit 4 Starter Packages, which combine the Qubit 4 Fluorometer with different assay kits for the quantification of nucleic acids or proteins. Qubit reagents bind selectively to DNA, RNA, or protein and only emit a fluorescent signal when bound to the target. This minimizes the effects of contaminants on the result and ensures accurate readings for sensitive samples.
With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result. Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds. Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.
Qubit 4 Quantitation Starter Kit
The Qubit 4 With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result. Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds. Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.
Qubit 4 Quantitation Starter Kit (Cat. No. Q33239) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit 4 System Verification Assay Kit, 50 assays (Cat. No. Q33237)
• Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33230)
• Qubit 1X dsDNA BR Assay Kit, 100 assays (Cat. No. Q33265)
• Qubit RNA IQ Assay Kit, 75 assays (Cat. No. Q33221)
• Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
• Qubit RNA BR Assay Kit, 100 assays (Cat. No. Q10210)
• Qubit Protein BR Assay Kit, 100 assays (Cat. No. A50668)
Qubit 4 NGS (Next-Generation Sequencing) Starter Kit
The Qubit NGS Starter Kit (Cat. No. Q33240) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Wi-Fi Dongle (Cat. No. A26774)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)
Qubit Protein BR Starter Kit
The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, and complex protein mixtures. The assay can deliver accurate protein quantification across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods.
The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use with the Qubit 4 Fluorometer. The assay is not designed for use with other Qubit models. The kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then, insert your sample into a Qubit 4 instrument and read the concentration.
The Qubit 4 Protein BR Starter Kit (Cat. No. A51292) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Wi-Fi Dongle (Cat. No. A26774)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit Protein BR Assay Kit, 100 assays (Cat. No. A50668)
Qubit 4 RNA IQ Starter Kit
The Qubit RNA IQ assay provides a fast, simple method to check whether an RNA sample has degraded. The assay utilizes two unique dyes: one that binds to large, intact RNA, and another that selectively binds to small, degraded RNA. Together they enable you to quickly assess the quality and integrity of an RNA sample. To use, simply add your samples to the RNA IQ working solution, then measure on the Qubit 4 Fluorometer.
The Qubit 4 RNA IQ Starter Package (Cat. No. Q33241) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit RNA IQ Assay Kit, 275 assays (Cat. No. Q33222)

Specifications
1 to 20 μL | |
470 nm (Blue LED), 635 nm (Red LED) | |
Qubit | |
sdDNA Quantification, RNA Quantification, Protein Quantification |
Fluorometer, Assay Tubes, Wi-Fi Dongle, Assay Kit | |
Qubit Fluorometer | |
Qubit 4 NGS Starter Kit, with WiFi | |
1 kit |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.