Learn More
DWK Life Sciences Crimp Cap, ND11
Crimp closures generally known for excellent sealing characteristics, even when it comes to long-term sample storage.
Brand:  DWK Life Sciences 225684
 View more versions of this product
Product Code. 16616872
  • £8.80 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The quality of a crimp seal is strongly dependent on the right calibration of the crimping tool used to apply the closure to the vial. A too tight or too loose crimp will impair the quality of the seal. To maintain good results, make sure to re-calibrate your crimping tool regularly and check the quality of the seal every time you crimp.
  • Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Silver
With liner
ND20
Red Rubber, Clear PTFE
Open Top Cap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Crimp Cap, ND11 > Red rubber, PTFE clear