missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Quick Gel Extraction and PCR Purification Combo Kit

Offers ability to perform both a gel extraction or PCR purification with single kit.

Brand:  Invitrogen™ K220001


Product Code. 10606845

  • £108.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: The PureLink Quick Gel Extraction & PCR Purification Combo Kit includes sufficient PureLink columns, collection tubes, elution tubes, and reagents for 50 preps.
This item is not returnable. View return policy

Description

Description

PureLink™ Quick Gel Extraction and PCR Purification Combo Kit offers the ability to perform both a gel extraction or a PCR purification a single kit.

The PureLink™ Quick Gel Extraction and PCR Purification Combo Kit is designed to purify DNA fragments from agarose gels. The simple procedure uses a silica-based spin cartridge to purify DNA fragments from 40 bp to 10 kb in < 30 minutes. It also provide all components to perform a rapid and efficient removal of short primers, dNTPs, enzymes, short-failed PCR products, and salts from PCR fragments >100 bp in less than 10 minutes. Isolated DNA using the PureLink™ kit is free of proteins, dye, and agarose, and is ready-to-use for a variety of applications, including automated fluorescent DNA sequencing, manual DNA sequencing, PCR, in vitro transcription, restriction mapping, cloning, and labeling. The kit contains all the buffers from the PureLink™ Quick Gel extraction kit and the PCR binding buffer from PureLink™ PCR Purification kit with Invitrogen branded lidded column.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Mini
50 μL
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
Ethidium Bromide, SYBR Safe
Room Temperature
15 μg (Binding capacity)
Silica Spin Column
Linear DNA fragments from: Agarose Gels, PCR Purification, Restriction Enzyme Reactions, PCR Products
DNA
Not High-throughput Compatible (Manual)
50 Preps
Up to 30 min.
Agarose Gel: ≤400 mg, PCR Product: ≤100 μL, or ≤40 μg dsDNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3130985 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberK220001
Lot Number2795605 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK220001
Lot Number2803762 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK220001
Lot Number2817749 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK220001
Lot Number2795608 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK220001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ PureLink™ Quick Gel Extraction and PCR Purification Combo Kit > Quantity: 50 Preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.