missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ 1M Tris-HCI, pH 8.0
GREENER_CHOICE

Pre-mixed, pH-adjusted, sterile-filtered solutions

Brand:  Invitrogen™ 15568025

Product Code. 10434742

  • £77.25 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Prepared as 1M concentrate, buffers can be diluted to desired concentration
  • Used in molecular biology or general biochemistry applications
  • Performance and Quality Testing: No DNase, RNase, or protease activity detected
  • Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Maxiprep, Midiprep, Miniprep, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 M
pH 8.0
Tris-HCl Buffer
Room Temperature
Store in refrigerator (2–8°C).
UltraPure™
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2392410 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15568025
Lot Number2451390 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15568025
Lot Number2451392 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15568025
Lot Number2451402 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15568025
Lot Number2488763 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15568025
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ UltraPure™ 1M Tris-HCI, pH 8.0 > Quantity: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.