Learn More
Invitrogen™ Yeast Homozygous Diploid Plate
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Brand:  Invitrogen™ 95401.H1P
Product Code. 10747245
  • £317.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yeast Homozygous Diploid Plate
Yeast deletion library created by collaboration of 8 North American and 8 European labs composing the Saccharomyces Genome Deletion Project
Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)
Individual Plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Yeast Homozygous Diploid Plate > Homozygous Diploid, Individual plate