missing translation for 'onlineSavingsMsg'
Learn More
Iodine, puriss., Honeywell Fluka™ Product Code.: 15630660

Iodine, puriss., Honeywell Fluka™
BIOPHARMA

Product Code. 15630660 Shop All Honeywell Fluka Products
6 x 1 kg
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
6 x 100 g
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15630660

Brand: Honeywell-Fluka 030026X1KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Quantity 6 x 1 kg
UN Number UN3495
IUPAC Name molecular iodine
Formula Weight 253.81
Percent Purity 99.8 to 100.5%
Grade puriss.
Vapor Pressure 0.31mmHg (25°C), 1mmHg (38.7°C)
Density Vapor Density: 9 (vs air)
Chemical Name or Material Iodine, Meets analytical specification of Ph. Eur.BP, USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine, puriss., Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.