missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, puriss., Honeywell Fluka™

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%
£34.19 - £1150.52
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15600660
|
Honeywell-Fluka
03002-25G |
25 g | N/A | N/A | N/A | |||||
15680650
![]() |
Honeywell-Fluka
03002-100G |
100 g |
£83.01
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15610660
![]() |
Honeywell-Fluka
03002-500G |
500 g |
£132.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15620660
|
Honeywell-Fluka
03002-6X100G |
6 x 100 g | N/A | N/A | N/A | |||||
15690650
![]() |
Honeywell-Fluka
03002-1KG |
1 kg |
£245.17
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15630660
![]() |
Honeywell-Fluka
03002-6X1KG |
6 x 1 kg |
£1150.52
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
7553-56-2 | |
MFCD00011355 MFCD00164163 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
253.81g/mol | |
puriss. | |
Vapor Density: 9 (vs air) |
I2 | |
3495 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
99.8 to 100.5% | |
0.31mmHg (25°C), 1mmHg (38.7°C) | |
Iodine, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling
P261-P273-P280-P305 + P351 + P338-P314
H312 + H332-H315-H319-H335-H372-H400
missing translation for 'einecsNumber' : 231-442-4
missing translation for 'rtecsNumber' : NN1575000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iodine, puriss., Honeywell Fluka™