Learn More

- Home
- Products
- Chemicals
- Nonmetals and Halogens
- Iodine (I)
- Iodine, puriss., Honeywell Fluka™
Iodine, puriss., Honeywell Fluka™
Product Code. 15600660 Shop All Honeywell Fluka ProductsProduct Code. 15600660
Brand: Honeywell-Fluka 0300225G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Chemical Identifiers
CAS | 7553-56-2 |
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Specifications
Quantity | 25 g |
UN Number | 3495 |
Beilstein | 3587194 |
IUPAC Name | molecular iodine |
Formula Weight | 253.81g/mol |
Percent Purity | 99.8 to 100.5% |
Grade | puriss. |
Vapor Pressure | 0.31mmHg (25°C), 1mmHg (38.7°C) |
Density | Vapor Density: 9 (vs air) |
Chemical Name or Material | Iodine, Meets analytical specification of Ph. Eur.BP, USP |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10300814
-
-
-
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 10244202
£155.00 / 5 L -
Catalog No. 10162180
£84.30 / 2.50 L -
Catalog No. 10598800
£140.00 / 2.50 L -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10784751
£287.00 / 2.50 L -
Catalog No. 13277983
£48.94 / 5 L -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10510781
£84.15 / 2.50 L -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10694722
£92.85 / 100g -
Catalog No. 10439750
£85.30 / 100g -
Catalog No. 10626782
£36.51 / 100g
Your input is important to us. Please complete this form to provide feedback related to the content on this product.