missing translation for 'onlineSavingsMsg'
Learn More
Iodine, resublimed crystals, ACS, 99.8%, Thermo Scientific Chemicals Product Code.: 11324947

Iodine, resublimed crystals, ACS, 99.8%, Thermo Scientific Chemicals

Product Code. 11324947
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11324947

Brand: Thermo Scientific Alfa Aesar 041955.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Melting Point 183°C to 186° (sublimation)
Color Gray-Purple
Physical Form Prills or Crystals
Quantity 500 g
Assay Percent Range 99.8%
Formula Weight 253.81
Grade ACS
Odor Irritating
Appearance Bluish-black
Density 4.93
Chemical Name or Material Iodine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L045 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number041955.22,041955.36
Lot NumberM10L007 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number041955.22,041955.36
Lot NumberT29K016 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number041955.22,041955.36
Lot NumberR03K035 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number041955.22,041955.36
Lot NumberW01J039 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number041955.22,041955.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine, resublimed crystals, ACS, 99.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.