missing translation for 'onlineSavingsMsg'
Learn More
Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11356428

Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11356428
10 g
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy

Product Code. 11356428

Brand: Thermo Scientific Alfa Aesar 012071.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The primary use of iridium is as a hardening agent in platinum alloys. Other uses are for the making of crucibles and devices that require high temperatures. Osmium/iridium alloys are used for compass bearings. Iridium is commonly used in complexes like Ir(mppy)3 and other complexes in polymer LED technology to increase the efficiency from 25% to about 100% due to triplet harvesting. Used in high-dose radiation therapy for the treatment of prostate and other forms of cancer. Iridium is used in tips of ballpoint pens. Iridium is used as a catalyst for carbonylation of methanol to create acetic acid. At one time, iridium as an alloy with platinum, was used in bushing the vents of heavy ordnance and, in a finely powdered condition (iridium black) for painting porcelain black.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Quantity 10 g
UN Number UN3089
Solubility Information Soluble in aqua regia.
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Iridium black

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012071.09,012071.04,012071.02
Lot NumberW23I033 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number012071.09,012071.04,012075.03,012071.02
Lot NumberW23G032 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012071.09,012071.04,012075.03,012071.02
Lot NumberI19M16 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012071.09,012071.04,012075.03,012071.02
Lot Number61200905 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012071.09,012071.04,012075.03,012071.02

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.