missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) chloride tetrahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11497797

Iron(II) chloride tetrahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11497797
250 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11497797

Brand: Thermo Scientific Alfa Aesar A16327.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iron(II) chloride tetrahydrate is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in waste water treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) chloride tetrahydrate is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in waste water treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

Solubility
Soluble in water.

Notes
Air and light-sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong base, strong acids, ethylene oxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.805
MDL Number MFCD00149709
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiiiShow MoreShow Less
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl

Specifications

Quantity 250 g
Linear Formula FeCl2·4H2O
UN Number UN3260
Merck Index 14,4043
Solubility Information Soluble in water.
Formula Weight 198.81 (126.75 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 1.93
Chemical Name or Material Iron(II) chloride tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252838 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberA16327.30,A16327.0B
Lot Number10252180 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog NumberA16327.30,A16327.0B
Lot NumberT29K035 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberA16327.30,A16327.0B
Lot NumberS10K044 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog NumberA16327.30,A16327.0B
Lot NumberQ10K026 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog NumberA16327.30,A16327.0B
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) chloride tetrahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.