missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10628144

Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10628144
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10628144

Brand: Thermo Scientific Acros 423731000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironateShow MoreShow Less
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64.0°C
Color Blue or Blue-Green or Green
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99+%
Linear Formula FeSO4·7H2O
Fieser 01,393; 04,237; 05,308
Merck Index 15,4084
Solubility Information Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Formula Weight 278.01
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464482 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number423730030,423730050,423730100,423735000,423731000
Lot NumberA0451946 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number423730030,423730050,423730100,423735000,423731000
Lot NumberA0458553 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number423730030,423730050,423730100,423735000,423731000
Lot NumberA0447318 Certificate TypeCertificate of Analysis Date01/12/2023 Catalog Number423730030,423730050,423730100,423735000,423731000
Lot NumberA0456214 Certificate TypeCertificate of Analysis Date08/09/2023 Catalog Number423730030,423730050,423730100,423735000,423731000
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.