Learn More
Iron(III) acetylacetonate, 99+%, Thermo Scientific Chemicals
£23.80 - £262.00
Chemical Identifiers
CAS | 14024-18-1 |
---|---|
Molecular Formula | C15H21FeO6 |
Molecular Weight (g/mol) | 353.17 |
MDL Number | MFCD00000020 |
InChI Key | AQBLLJNPHDIAPN-LNTINUHCSA-K |
Synonym | 2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate |
IUPAC Name | iron(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
SMILES | [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10624772
|
Thermo Scientific Acros
119130250 |
25 g | Glass bottle |
£23.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061000
|
Thermo Scientific Acros
119131000 |
100 g | Glass bottle |
£45.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540751
|
Thermo Scientific Acros
119132500 |
250 g | Glass bottle |
£88.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15169173
|
Thermo Scientific Acros
119130010 |
1 kg | Glass bottle |
£262.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14024-18-1 | |
353.17 | |
AQBLLJNPHDIAPN-LNTINUHCSA-K | |
iron(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
C15H21FeO6 | |
MFCD00000020 | |
2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate | |
[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
14024-18-1 | |
Red | |
99% min. (Iodometry) | |
MFCD00000020 | |
2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate | |
AQBLLJNPHDIAPN-LNTINUHCSA-K | |
iron(3+) tris((2Z)-4-oxopent-2-en-2-olate) | |
353.17 | |
Powder |
180.0°C to 183.0°C | |
Authentic | |
C15H21FeO6 | |
04,268; 06,304; 13,211 | |
Solubility in water: 2g/L (20°C). Other solubilities: readily soluble in acetone and chloroform,sparingly soluble in ether,40.1g/kg alcohol (20°C),535.4g/kg benzene (20°C) | |
[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
353.17 | |
99+% | |
Iron(III) acetylacetonate, 99+% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 237-853-5
missing translation for 'rtecsNumber' : NO8960000
missing translation for 'tsca' : TSCA
- Iron(III) acetylacetonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only