missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals Product Code.: 10306741

Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals

Product Code. 10306741
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10306741

Brand: Thermo Scientific Acros 345230050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142906-29-4
Molecular Formula Fe2O12S3·5H2O
Molecular Weight (g/mol) 489.96
MDL Number MFCD00149714
InChI Key YHGPYBQVSJBGHH-UHFFFAOYSA-H
Synonym unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrateShow MoreShow Less
PubChem CID 23443659
IUPAC Name iron(3+);trisulfate;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Color Beige-Yellow to White
Physical Form Crystalline or Granular Powder
Quantity 5 g
Assay Percent Range 97%
Linear Formula Fe2(SO4)3·5H2O
Solubility Information Solubility in water: soluble
Formula Weight 489.96
Percent Purity 97%
Packaging Glass Bottle
Chemical Name or Material Iron(III) sulfate pentahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466593 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog Number345230051,345230050,345235000,345231000
Lot NumberA0465398 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog Number345230051,345230050,345235000,345231000
Lot NumberA0463423 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number345230051,345230050,345235000,345231000
Lot NumberA0461464 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number345230051,345230050,345235000,345231000
Lot NumberA0459459 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number345230051,345230050,345235000,345231000
compliance-icons compliance-icons
Product Identifier
  • Iron(III)sulfate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.