missing translation for 'onlineSavingsMsg'
Learn More

Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 036683.22

62.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11314207

  • £62.31 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Iron Nickel powder
Silver gray powder
Ni:Fe; 50:50 wt%
100 g
-325 Mesh
Powder
Silver to Gray
>99.5% (metals basis)
MFCD01741928
Insoluble in water.
Odorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU14A023 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036683.14,036683.36,036683.22
Lot NumberS17F021 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036683.14,036683.36,036683.22
Lot NumberB11P17 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036683.14,036683.36,036683.22
Lot NumberW10B037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036683.14,036683.36,036683.22
Lot NumberE20H23 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036683.14,036683.36,036683.22
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals > 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only