Learn More
Iron Nickel powder, -325 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
£33.67 - £172.86
Specifications
Chemical Name or Material | Iron Nickel powder |
---|---|
Color | Silver to Gray |
Appearance | Silver gray powder |
Assay Percent Range | >99.5% (metals basis) |
Molecular Formula | Ni:Fe; 50:50 wt% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466165
|
Thermo Scientific Alfa Aesar
036683.14 |
25 g |
£33.67
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314207
|
Thermo Scientific Alfa Aesar
036683.22 |
100 g |
£62.31
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324207
|
Thermo Scientific Alfa Aesar
036683.36 |
500 g |
£172.86
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron Nickel powder | |
Silver gray powder | |
Ni:Fe; 50:50 wt% | |
Insoluble in water. | |
Odorless |
Silver to Gray | |
>99.5% (metals basis) | |
MFCD01741928 | |
-325 Mesh | |
Powder |
Safety and Handling
Keep container tightly closed. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
H228-H317-H350-H372
missing translation for 'dotInformation' : DOT Class: 4.1; Packing Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Iron Nickel powder
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Nickel, Iron
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.