missing translation for 'onlineSavingsMsg'
Learn More
Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™ Product Code.: 10409360

Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™

Product Code. 10409360
500 mL, Amber Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
500 mL
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10409360

Brand: Fisher Chemical A/7000/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
MDL Number 2934

Specifications

Melting Point -117°C
pH 6.5
Boiling Point 130°C
Packaging Amber Glass Bottle
Molecular Formula C5H12O
Quantity 500 mL
Vapor Density 3.04
Formula Weight 88.15g/mol
Vapor Pressure 4 hPa at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • iso-Amyl alcohol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.