missing translation for 'onlineSavingsMsg'
Learn More
Isobutyryl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11405947

Isobutyryl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11405947
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11405947

Brand: Thermo Scientific Alfa Aesar B24472.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

Solubility
Miscible with chloroform, glacial acetic acid, ether, toluene, methylene chloride and benzene.

Notes
Moisture sensitive. Incompatible with bases, water, strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-30-1
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00000717
InChI Key DGMOBVGABMBZSB-UHFFFAOYSA-N
Synonym isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chlorideShow MoreShow Less
PubChem CID 62325
IUPAC Name 2-methylpropanoyl chloride
SMILES CC(C)C(=O)Cl

Specifications

Melting Point ∼-90°C
Density 1.017
Boiling Point 91°C to 93°C
Flash Point 8°C (46°F)
Assay Percent Range 0.98
Odor Unpleasant
Refractive Index 1.407
Quantity 500 g
UN Number UN2395
Beilstein 605393
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform,glacial acetic acid,ether,toluene,methylene chloride and benzene.
Formula Weight 106.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyryl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberGBFA008353 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24472.36,B24472.22
Lot Number10163479 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24472.36,B24472.22
Lot Number10236169 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24472.36,B24472.22
Lot Number10140374 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24472.36,B24472.22
Lot Number10132254 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24472.36,B24472.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutyryl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.