missing translation for 'onlineSavingsMsg'
Learn More
Isophthalic acid, 99%, Thermo Scientific Chemicals Product Code.: 10755761

Isophthalic acid, 99%, Thermo Scientific Chemicals

Product Code. 10755761
2 kg, Plastic bottle
Click to view available options
Quantity:
5 g
500 g
2 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10755761

Brand: Thermo Scientific Acros 122650020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fjShow MoreShow Less
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 341.0°C to 343.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H4(CO2H)2
Quantity 2 kg
Beilstein 09, 832
Merck Index 15, 5243
Solubility Information Solubility in water: 0.01g/100mL (25 c). Other solubilities: freely soluble in alcohol and glacial acetic acid,practically insoluble in benzene and petroleum,ether
Formula Weight 166.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Isophthalic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466856 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number122650020,122655000
Lot NumberA0435619 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog Number122650020,122655000
Lot NumberA0455513 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number122650050,122650020,122655000
Lot NumberA0447067 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number122650050,122650020,122655000
Lot NumberA0447068 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number122650050,122650020,122655000
Product Identifier
  • Isophthalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isophthalic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.