missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free Product Code.: 10205731

Thermo Scientific Chemicals Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free

Product Code. 10205731
10 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10205731

Brand: Thermo Scientific Chemicals 302790100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactosideShow MoreShow Less
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Beilstein 17, IV, 3726
Merck Index 15, 5126
Specific Rotation Condition − 31.00 (20.00°C c=1,water)
Specific Rotation -29° to -33° (20°C, 589nm) (c=1, water)
Formula Weight 238.31
Percent Purity ≥98.5%
Grade Biochemical
Physical Form Powder or Flakes
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459528 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number302790010,302790250,302790100
Lot NumberA0456255 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number302790010,302790250,302790100
Lot NumberA0449495 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number302790010,302790250,302790100
Lot NumberA0415080 Certificate TypeCertificate of Analysis Date24/06/2022 Catalog Number302790010,302790250,302790100
Lot NumberA0410837 Certificate TypeCertificate of Analysis Date28/01/2022 Catalog Number302790010,302790250,302790100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.