missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaolin, pure, Thermo Scientific Chemicals
£29.70 - £139.00
Chemical Identifiers
CAS | 1332-58-7 |
---|---|
Molecular Formula | Al2H4O9Si2 |
Molecular Weight (g/mol) | 258.16 |
MDL Number | MFCD00062311 |
InChI Key | NLYAJNPCOHFWQQ-UHFFFAOYSA-N |
Synonym | kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss |
PubChem CID | 56841936 |
IUPAC Name | oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate |
SMILES | O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772441
|
Thermo Scientific Acros
211740010 |
1 kg | Plastic Bottle |
£29.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752574
|
Thermo Scientific Acros
211740025 |
2.5 kg | Plastic Bottle |
£51.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984321
|
Thermo Scientific Acros
211740100 |
10 kg | Plastic drum |
£139.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1332-58-7 | |
258.16 | |
NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
56841936 | |
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O |
Al2H4O9Si2 | |
MFCD00062311 | |
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss | |
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate |
Specifications
1750.0°C | |
Beige | |
Al2H4O9Si2 | |
14,5282 | |
Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkali hydroxides | |
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O | |
258.16 | |
258.2 | |
1.5% max. | |
Kaolin |
1332-58-7 | |
Powder | |
MFCD00062311 | |
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss | |
NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate | |
56841936 | |
Pure | |
10.8% to 11.8% |
RUO – Research Use Only