Learn More
SI Analytics™ Karl-Fischer™ Titration Vessel
SI Analytics™ Karl-Fischer™ Titration Vessel is made of glass and get suited for coulometry.
Brand: SI Analytics™ 285216630
Description
- Constructed of glass
- Suitable for coulometry
- Includes one NS 19 opening and one NS 7.5 opening
- GL 18 thread size
Specifications
Titration Vessel | |
Openings | |
1 x NS 19, 1 x NS 7.5 |
Coulometry | |
Glass |