missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A potent inhibitor of inositol monophophatase; stable to hydrolysis. Induces autophagy in COS-7 cells independently of mTOR inhibition.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 298.13 |
Percent Purity | >98% |
Chemical Name or Material | L-690, 330 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-690,330, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction